(6-Methyl-3-pyridinyl)methanol ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
6-Methyl-3-pyridinemethanol, 5-Hydroxymethyl-2-methylpyridine, (6-Methylpyridin-3-yl)methanol, 5-Hydroxymethyl-2-picoline
M2683-1G
M2683-5G
TCM2683-1GEA
56.25
CAD
TCM2683-1G
TCM2683-5G
(6-Methyl-3-pyridinyl)methanol ≥97.0% (by GC, titration analysis)
(6-Methyl-3-pyridinyl)methanol
Formula:
C₇H₉NO Boiling Pt: 100°C |
MDL Number:
MFCD08692003 CAS Number: 34107-46-5 |
Specification Test Results
Appearance | Light yellow to Brown powder to lump |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 44.0 to 48.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...