1-(2-Hydroxyphenyl)-3-(4-methoxyphenyl)-2-propen-1-one ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

H1484-1G H1484-5G
TCH1484-1GEA 129.56 CAD
TCH1484-1G TCH1484-5G
1-(2-Hydroxyphenyl)-3-(4-methoxyphenyl)-2-propen-1-one ≥98.0% (by HPLC, titration analysis)
1-(2-Hydroxyphenyl)-3-(4-methoxyphenyl)-2-propen-1-one
Formula: C₁₆H₁₄O₃
MDL Number: MFCD00016447
CAS Number: 3327-24-0

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 91.0 to 95.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR