tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate ≥97.0% (by GC)

Supplier: TCI America

B5501-1G B5501-5G
TCB5501-1GEA 225.94 CAD
TCB5501-1G TCB5501-5G
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate ≥97.0% (by GC)
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate
Formula: C₁₁H₁₉NO₃
MDL Number: MFCD07779385
CAS Number: 147804-30-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 55.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR