Methyl thieno[3,2-b]thiophene-2-carboxylate ≥98.0% (by GC)

Supplier: TCI America

M3001-1G M3001-5G
TCM3001-1GEA 210.14 CAD
TCM3001-1G TCM3001-5G
Methyl thieno[3,2-b]thiophene-2-carboxylate ≥98.0% (by GC)
Methyl thieno[3,2-b]thiophene-2-carboxylate
Formula: C₈H₆O₂S₂
MDL Number: MFCD01934498
CAS Number: 98800-10-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 100.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR