1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid, 1-(Ethoxycarbonyl)piperidine-4-carboxylic acid

E1208-1G E1208-5G
76220-030EA 56.88 CAD
76220-030 76220-032
1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid ≥98.0% (by GC, titration analysis)
1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid
Formula: C₉H₁₅NO₄
MW: 201.22 g/mol
MDL Number: MFCD07339354
CAS Number: 118133-15-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR