Ethyl-5-amino-1-benzofuran-2-carboxylate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E1138-1G E1138-5G
TCE1138-1GEA 107.44 CAD
TCE1138-1G TCE1138-5G
Ethyl-5-amino-1-benzofuran-2-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl-5-amino-1-benzofuran-2-carboxylate
Formula: C₁₁H₁₁NO₃
MDL Number: MFCD08275091
CAS Number: 174775-48-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 62.0 to 66.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR