2-Bromo-4-methoxy-6-nitrobenzenol ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Bromo-4-methoxy-6-nitrophenol

B4723-1G B4723-5G
TCB4723-1GEA 58.46 CAD
TCB4723-1G TCB4723-5G
2-Bromo-4-methoxy-6-nitrobenzenol ≥98.0% (by GC)
2-Bromo-4-methoxy-6-nitrobenzenol
Formula: C₇H₆BrNO₄
MW: 248.04 g/mol
MDL Number: MFCD06797984
CAS Number: 115929-59-4

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 116.0 to 120.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR