Methyl-3-hydroxy-2-thiophenecarboxylate ≥97.0% (by GC)

Supplier: TCI America

M2716-1G M2716-5G
TCM2716-1GEA 47.4 CAD
TCM2716-1G TCM2716-5G
Methyl-3-hydroxy-2-thiophenecarboxylate ≥97.0% (by GC)
Methyl-3-hydroxy-2-thiophenecarboxylate
Formula: C₆H₆O₃S
Boiling Pt: 108°C
MDL Number: MFCD00055642
CAS Number: 5118-06-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 43.0 to 47.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR