6-(2-Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one 98,0 GC_ASSAY_METHOD
Supplier: TCI America
|
Warning
|
Synonyms:
6-(Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one, 6-(2-Chloroacetyl)-2H-benzo[b][1,4]oxazin-3(4H)-one
C3097-25G
C3097-5G
TCC3097-25GEA
297.04
CAD
TCC3097-25G
TCC3097-5G
6-(2-Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one 98,0 GC_ASSAY_METHOD
6-(2-Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one
Formula:
C₁₀H₈ClNO₃ MW: 225.63 g/mol |
MDL Number:
MFCD01321312 CAS Number: 26518-76-3 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 228.0 to 232.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...