2-(Hydroxymethyl)-4-nitrobenzenol ≥98.0% (by GC)

Supplier: TCI America

H1549-25G H1549-5G
TCH1549-25GEA 402.9 CAD
TCH1549-25G TCH1549-5G
2-(Hydroxymethyl)-4-nitrobenzenol ≥98.0% (by GC)
2-(Hydroxymethyl)-4-nitrobenzenol
Formula: C₇H₇NO₄
MDL Number: MFCD01321361
CAS Number: 39224-61-8

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 123.0 to 127.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR