5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

H1550-1G H1550-200MG
TCH1550-1GEA 153.73 CAD
TCH1550-1G TCH1550-200MG
5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde ≥98.0% (by GC)
5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde
Formula: C₁₀H₁₃NO₂S
MDL Number: MFCD03425728
CAS Number: 207290-72-0

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 143.0 to 147.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR