3-(Dimethylamino)-1-phenyl-2-propen-1-one ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D5384-1G D5384-5G
TCD5384-1GEA 104.28 CAD
TCD5384-1G TCD5384-5G
3-(Dimethylamino)-1-phenyl-2-propen-1-one ≥98.0% (by GC, titration analysis)
3-(Dimethylamino)-1-phenyl-2-propen-1-one
Formula: C₁₁H₁₃NO
MDL Number: MFCD00121193
CAS Number: 1201-93-0

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 90.0 to 94.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR