Ethyl-6-methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid ethyl ester, Ethyl 6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylate

E1131-1G E1131-200MG
TCE1131-1GEA 629.79 CAD
TCE1131-1G TCE1131-200MG
Ethyl-6-methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥98.0% (by HPLC, total nitrogen)
Ethyl-6-methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate
Formula: C₁₄H₁₆N₂O₃
MW: 260.3 g/mol
MDL Number: MFCD00124778
CAS Number: 5395-36-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 206.0 to 210.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR