Methyl-1,5-dimethyl-1H-pyrazole-3-carboxylate ≥98.0% (by GC)

Supplier: TCI America
Warning

M2863-1G M2863-5G
TCM2863-1GEA 178.54 CAD
TCM2863-1G TCM2863-5G
Methyl-1,5-dimethyl-1H-pyrazole-3-carboxylate ≥98.0% (by GC)
Methyl-1,5-dimethyl-1H-pyrazole-3-carboxylate
Formula: C₇H₁₀N₂O₂
Boiling Pt: 144°C
MDL Number: MFCD06661312
CAS Number: 10250-61-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 73.0 to 77.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR