4,4,4-Trifluoro-1-(4-methoxyphenyl)butane-1,3-dione ≥98.0% (by GC)

Supplier: TCI America

T3283-1G T3283-5G
TCT3283-1GEA 137.46 CAD
TCT3283-1G TCT3283-5G
4,4,4-Trifluoro-1-(4-methoxyphenyl)butane-1,3-dione ≥98.0% (by GC)
4,4,4-Trifluoro-1-(4-methoxyphenyl)butane-1,3-dione
Formula: C₁₁H₉F₃O₃
MDL Number: MFCD03238723
CAS Number: 15191-68-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR