1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione 98,0 GC, T

Supplier: TCI America

Synonyms: 1-(4-Chlorophenyl)-4,4,4-trifluorobutane-1,3-dione, 3-(4-Chlorobenzoyl)-1,1,1-trifluoroacetone

C3139-1G C3139-5G
76218-906EA 66.36 CAD
76218-906 76218-908
1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione 98,0 GC, T
1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione
Formula: C₁₀H₆ClF₃O₂
MW: 250.61 g/mol
MDL Number: MFCD00511276
CAS Number: 18931-60-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 60.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR