4-Amino-5-(ethylsulfonyl)-2-methoxybenzoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A2615-1G
A2615-5G
TCA2615-1GEA
50.56
CAD
TCA2615-1G
TCA2615-5G
4-Amino-5-(ethylsulfonyl)-2-methoxybenzoic acid ≥98.0% (by HPLC, titration analysis)
4-Amino-5-(ethylsulfonyl)-2-methoxybenzoic acid
Formula:
C₁₀H₁₃NO₅S |
MDL Number:
MFCD04973619 CAS Number: 71675-87-1 EINECS: 275-833-8 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystalline |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 148.0 to 152.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...