2,4-Difluoro-3-cyanophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
C3226-1G
C3226-200MG
TCC3226-1GEA
192.76
CAD
TCC3226-1G
TCC3226-200MG
2,4-Difluoro-3-cyanophenylboronic acid (contains varying amounts of Anhydride)
2,4-Difluoro-3-cyanophenylboronic acid
Formula:
C₇H₄BF₂NO₂ Melting Pt: 134°C |
MDL Number:
MFCD12026699 CAS Number: 871940-31-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 111.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...