5,11-Dihydroindolo[3,2-b]carbazole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: Indolo[3,2-b]carbazole

D5124-1G D5124-200MG
TCD5124-1GEA 504.02 CAD
TCD5124-1G TCD5124-200MG
5,11-Dihydroindolo[3,2-b]carbazole ≥98.0% (by HPLC, total nitrogen)
5,11-Dihydroindolo[3,2-b]carbazole
Formula: C₁₈H₁₂N₂
Melting Pt: 460°C
MDL Number: MFCD09879258
CAS Number: 6336-32-9

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR