2-(2-Propynyloxy)benzenecarbaldehyde ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
P2338-1G
P2338-200MG
76220-410EA
399.74
CAD
76220-410
76220-412
2-(2-Propynyloxy)benzenecarbaldehyde ≥98.0% (by GC)
2-(2-Propynyloxy)benzenecarbaldehyde
Formula:
C₁₀H₈O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD02380378 CAS Number: 29978-83-4 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 68.0 to 72.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...