5-Bromo-1H-pyrrolo[2,3-b]pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B3588-1G B3588-5G
TCB3588-1GEA 56.88 CAD
TCB3588-1G TCB3588-5G
5-Bromo-1H-pyrrolo[2,3-b]pyridine ≥98.0% (by GC, titration analysis)
5-Bromo-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₅BrN₂
MDL Number: MFCD06659677
CAS Number: 183208-35-7

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 178.0 to 182.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR