Ethyl-5-fluoro-1H-indole-2-carboxylate ≥98.0% (by GC)

Supplier: TCI America
Warning

E1035-1G E1035-5G
TCE1035-1GEA 213.3 CAD
TCE1035-1G TCE1035-5G
Ethyl-5-fluoro-1H-indole-2-carboxylate ≥98.0% (by GC)
Ethyl-5-fluoro-1H-indole-2-carboxylate
Formula: C₁₁H₁₀FNO₂
MDL Number: MFCD00152076
CAS Number: 348-36-7

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR