5-Fluoro-3-methylindole ≥98.0% (by GC)

Supplier: TCI America
Warning

F1053-1G F1053-200MG
TCF1053-1GEA 293.88 CAD
TCF1053-1G TCF1053-200MG
5-Fluoro-3-methylindole ≥98.0% (by GC)
5-Fluoro-3-methylindole
Formula: C₉H₈FN
MDL Number: MFCD03095377
CAS Number: 392-13-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR