5,6-Diamino-1-methylpyrimidine-2,4(1H,3H)-dione ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 5,6-Diamino-1-methyluracil

D5010-1G D5010-5G
TCD5010-1GEA 64.78 CAD
TCD5010-1G TCD5010-5G
5,6-Diamino-1-methylpyrimidine-2,4(1H,3H)-dione ≥98.0% (by HPLC, total nitrogen)
5,6-Diamino-1-methylpyrimidine-2,4(1H,3H)-dione
Formula: C₅H₈N₄O₂
MW: 156.15 g/mol
Melting Pt: 270°C
MDL Number: MFCD00091924
CAS Number: 6972-82-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR