Methyl pipecolinate hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Methyl Pipecolinate HCl, H-DL-Pip-OMe·HCl, Methyl piperidine-2-carboxylate hydrochloride, Methyl 2-piperidinecarboxylate hydrochloride, Pipecolinic acid methyl ester hydrochloride, 2-Piperidinecarboxylic acid methyl ester hydrochloride
M2591-1G
M2591-5G
TCM2591-1GEA
64.78
CAD
TCM2591-1G
TCM2591-5G
Methyl pipecolinate hydrochloride ≥98.0% (by titrimetric analysis)
Methyl pipecolinate hydrochloride
Formula:
C₇H₁₄ClNO₂ MW: 179.64 g/mol |
MDL Number:
MFCD00192316 CAS Number: 32559-18-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...