4-(2-Morpholin-4-ylethoxy)phenylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M2691-1G M2691-5G
TCM2691-1GEA 60.04 CAD
TCM2691-1G TCM2691-5G
4-(2-Morpholin-4-ylethoxy)phenylamine ≥98.0% (by GC, titration analysis)
4-(2-Morpholin-4-ylethoxy)phenylamine
Formula: C₁₂H₁₈N₂O₂
Boiling Pt: 150°C
MDL Number: MFCD04970981
CAS Number: 52481-41-1

Order Now

Specification Test Results

Appearance Light yellow to Yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR