2,6-Dibromobenzenecarbaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2,6-Dibromobenzaldehyde

D4552-1G D4552-5G
TCD4552-1GEA 62.88 CAD
TCD4552-1G TCD4552-5G
2,6-Dibromobenzenecarbaldehyde ≥98.0% (by GC)
2,6-Dibromobenzenecarbaldehyde
Formula: C₇H₄Br₂O
MDL Number: MFCD11040410
CAS Number: 67713-23-9

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 90.0 to 94.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR