1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-one ≥98.0% (by GC)

Supplier: TCI America

D5069-1G D5069-200MG
TCD5069-1GEA 429.76 CAD
TCD5069-1G TCD5069-200MG
1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-one ≥98.0% (by GC)
1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-one
Formula: C₁₁H₁₀O₂
MDL Number: MFCD09955085
CAS Number: 196597-78-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 133.0 to 137.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR