1-(4-Propoxyphenyl)ethanone ≥98.0% (by GC)

Supplier: TCI America

P2377-1G P2377-5G
TCP2377-1GEA 107.44 CAD
TCP2377-1G TCP2377-5G
1-(4-Propoxyphenyl)ethanone ≥98.0% (by GC)
1-(4-Propoxyphenyl)ethanone
Formula: C₁₁H₁₄O₂
Boiling Pt: 111°C
Storage Temperature: Refrigerator
MDL Number: MFCD01922144
CAS Number: 5736-86-7

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR