5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A3038-1G A3038-5G
TCA3038-1GEA 52.14 CAD
TCA3038-1G TCA3038-5G
5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine ≥98.0% (by GC, titration analysis)
5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine
Formula: C₇H₆N₄S
MDL Number: MFCD00473692
CAS Number: 2002-04-2

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR