2,2-Difluoroethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

D4758-1G D4758-5G
TCD4758-1GEA 28.44 CAD
TCD4758-1G TCD4758-5G
2,2-Difluoroethylamine ≥98.0% (by GC, titration analysis)
2,2-Difluoroethylamine
Formula: C₂H₅F₂N
Boiling Pt: 68°C
MDL Number: MFCD00798137
CAS Number: 430-67-1
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR