1-(4-Chlorobenzyl)-1H-indole-3-carbaldehyde ≥95.0% (by HPLC)

Supplier: TCI America
Warning

O0450-10MG O0450-50MG
TCO0450-10MGEA 167.01 CAD
TCO0450-10MG TCO0450-50MG
1-(4-Chlorobenzyl)-1H-indole-3-carbaldehyde ≥95.0% (by HPLC)
1-(4-Chlorobenzyl)-1H-indole-3-carbaldehyde
Formula: C₁₆H₁₂ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD01051808
CAS Number: 75629-57-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 4.90 to 5.50 %
Melting point 115.0 to 119.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR