6-Bromo-2-oxo-2H-chromene-3-carboxylic acid ≥98.0% (by HPLC)

Supplier: TCI America
Warning

B4696-25G B4696-5G
TCB4696-25GEA 603.56 CAD
TCB4696-25G TCB4696-5G
6-Bromo-2-oxo-2H-chromene-3-carboxylic acid ≥98.0% (by HPLC)
6-Bromo-2-oxo-2H-chromene-3-carboxylic acid
Formula: C₁₀H₅BrO₄
Melting Pt: 193°C
MDL Number: MFCD00047640
CAS Number: 2199-87-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR