3-Carbamoylpyrrolidine-1-carboxylic acid tert-butyl ester ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B5248-1G B5248-5G
TCB5248-1GEA 189.6 CAD
TCB5248-1G TCB5248-5G
3-Carbamoylpyrrolidine-1-carboxylic acid tert-butyl ester ≥98.0% (by HPLC, total nitrogen)
3-Carbamoylpyrrolidine-1-carboxylic acid tert-butyl ester
Formula: C₁₀H₁₈N₂O₃
MDL Number: MFCD04116210
CAS Number: 122684-34-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR