2,2-Dimethyl-N-pyridin-4-ylpropionamide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,2-Dimethyl-N-pyridin-4-yl-propionamide, 2,2-Dimethyl-N-(4-pyridinyl)propanamide, N-(4-Pyridyl)pivalamide, 4-(2,2,2-Trimethylacetamido)pyridine, 2,2-Dimethyl-N-(4-pyridyl)propionamide

P2044-1G P2044-5G
TCP2044-1GEA 155.31 CAD
TCP2044-1G TCP2044-5G
2,2-Dimethyl-N-pyridin-4-ylpropionamide ≥98.0% (by GC)
2,2-Dimethyl-N-pyridin-4-ylpropionamide
Formula: C₁₀H₁₄N₂O
MW: 178.24 g/mol
MDL Number: MFCD00996248
CAS Number: 70298-89-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 131.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR