7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

C2897-1G C2897-5G
TCC2897-1GEA 71.1 CAD
TCC2897-1G TCC2897-5G
7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid ≥98.0% (by HPLC, titration analysis)
7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid
Formula: C₁₂H₉ClFNO₃
MDL Number: MFCD00792459
CAS Number: 68077-26-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR