5,6-Dichloro-1H-1,3-benzimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D5146-1G D5146-5G
TCD5146-1GEA 314.42 CAD
TCD5146-1G TCD5146-5G
5,6-Dichloro-1H-1,3-benzimidazole ≥98.0% (by GC, titration analysis)
5,6-Dichloro-1H-1,3-benzimidazole
Formula: C₇H₄Cl₂N₂
MDL Number: MFCD00211325
CAS Number: 6478-73-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 208.0 to 212.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR