1H,1H,7H-Perfluoroheptyl methacrylate ≥97.0% (by GC) stabilised

Supplier: TCI America
Warning

Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl Methacrylate, Methacrylic acid 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl ester

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

D4990-25G D4990-5G
76219-092EA 189.6 CAD
76219-092 76219-094
1H,1H,7H-Perfluoroheptyl methacrylate ≥97.0% (by GC) stabilised
1H,1H,7H-Perfluoroheptyl methacrylate
Formula: C₁₁H₈F₁₂O₂
MW: 400.17 g/mol
Boiling Pt: 112°C
Flash Pt: 104°C
Storage Temperature: Refrigerator
MDL Number: MFCD00080658
CAS Number: 2261-99-6
EINECS: 218-863-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR