1H,1H,2H,2H-Perfluorooctyl methacrylate ≥98.0% (by GC) stabilized

Supplier: TCI America
Warning


Stabilization: Stabilized with HQ + MEHQ

T3414-100G T3414-25G T3414-5G
76220-432EA 859.52 CAD
76220-432 76220-434 76220-436
1H,1H,2H,2H-Perfluorooctyl methacrylate ≥98.0% (by GC) stabilized
1H,1H,2H,2H-Perfluorooctyl methacrylate
Formula: C₁₂H₉F₁₃O₂
MDL Number: MFCD00077580
CAS Number: 2144-53-8
EINECS: 218-407-9

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR