4-Methylumbelliferyl-Α-L-arabinopyranoside ≥98.0% (by HPLC)
Supplier: TCI America
M3025-250MG
M3025-50MG
TCM3025-250MGEA
407.64
CAD
TCM3025-250MG
TCM3025-50MG
4-Methylumbelliferyl-Α-L-arabinopyranoside ≥98.0% (by HPLC)
4-Methylumbelliferyl-Α-L-arabinopyranoside
Formula:
C₁₅H₁₆O₇ Storage Temperature: Freezer |
MDL Number:
MFCD00064087 CAS Number: 69414-26-2 EINECS: 273-991-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Carbon) | 57.50 to 59.50 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...