Flavokawain A ≥98.0% (by GC)

Supplier: TCI America
Warning

H1571-1G H1571-5G
TCH1571-1GEA 101.12 CAD
TCH1571-1G TCH1571-5G
Flavokawain A ≥98.0% (by GC)
Flavokawain A
Formula: C₁₈H₁₈O₅
MDL Number: MFCD00017174
CAS Number: 3420-72-2

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 112.0 to 116.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR