2'-Hydroxy-2-phenylacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Benzyl 2-Hydroxyphenyl ketone, 2'-Hydroxy-2-phenylacetophenone, 1-(2-Hydroxyphenyl)-2-phenylethanone

B4673-1G B4673-200MG
TCB4673-1GEA 328.64 CAD
TCB4673-1G TCB4673-200MG
2'-Hydroxy-2-phenylacetophenone ≥98.0% (by GC)
2'-Hydroxy-2-phenylacetophenone
Formula: C₁₄H₁₂O₂
MW: 212.24 g/mol
Boiling Pt: 165°C
MDL Number: MFCD00218589
CAS Number: 2491-31-8

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR