(-)-Epigallocatechin ≥98.0% (by GC)

Supplier: TCI America

E1084-10MG E1084-50MG
TCE1084-10MGEA 216.46 CAD
TCE1084-10MG TCE1084-50MG
(-)-Epigallocatechin ≥98.0% (by GC)
(-)-Epigallocatechin
Formula: C₁₅H₁₄O₇
Storage Temperature: Refrigerator
MDL Number: MFCD00075939
CAS Number: 970-74-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -48.0 to -51.0 deg (C=0.5, Acetone)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR