6-Acetyl-2-Benzoxazolinone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 6-Acetyl-2-benzoxazolinone, 6-Acetyl-2(3H)-benzoxazolone, 6-Acetyl-2-hydroxybenzoxazole, 6-Acetyl-2-benzoxazolone, 6-Acetylbenzo[d]oxazol-2(3H)-one

A2907-1G A2907-5G
TCA2907-1GEA 110.6 CAD
TCA2907-1G TCA2907-5G
6-Acetyl-2-Benzoxazolinone ≥98.0% (by HPLC, titration analysis)
6-Acetyl-2-Benzoxazolinone
Formula: C₉H₇NO₃
MW: 177.16 g/mol
MDL Number: MFCD01664312
CAS Number: 54903-09-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 234.0 to 238.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR