4-Methylumbelliferyl-N-acetyl-β-D-galactosaminide hydrate ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
4-Methylumbelliferyl N-Acetyl-beta-D-galactosaminide, 4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-galactopyranoside
M3029-1G
M3029-250MG
TCM3029-1GEA
916.4
CAD
TCM3029-1G
TCM3029-250MG
4-Methylumbelliferyl-N-acetyl-β-D-galactosaminide hydrate ≥98.0% (by HPLC)
4-Methylumbelliferyl-N-acetyl-beta-D-galactosaminide hydrate
Formula:
C₁₈H₂₁NO₈ MW: 379.365 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00067658 CAS Number: 36476-29-6 EINECS: 253-052-3 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 3.40 to 4.00 % |
Specific rotation [a]20/D | +23.0 to +26.0 deg (C=0.4, DMSO) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...