(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
B5613-1G
B5613-5G
76218-862EA
162.74
CAD
76218-862
76218-864
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride ≥98.0% (by HPLC)
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride
Formula:
C₃₆H₅₂ClMnN₂O₂ Melting Pt: 326°C |
MDL Number:
MFCD02101664 CAS Number: 138124-32-0 Merck Index: 14,05252 |
Specification Test Results
Appearance | Orange to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Chelometric Titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...