Doxepin hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

D4626-1G D4626-25G D4626-5G
TCD4626-1GEA 93.22 CAD
TCD4626-1G TCD4626-25G TCD4626-5G
Doxepin hydrochloride ≥98.0% (by HPLC, titration analysis)
Formula: C₁₉H₂₂ClNO
MDL Number: MFCD00079135
CAS Number: 1229-29-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,03435

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 % (total of isomers)
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 186.0 to 190.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR