Zinc meso-tetraphenylporphine ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: 5,10,15,20-Tetraphenyl-21H,23H-porphine Zinc(II), Zinc(II) Tetraphenylporphyrin, 5,10,15,20-Tetraphenyl-21H,23H-porphine zinc, Zinc TPP, Zinc(II) TPP, Tetraphenylporphine zinc, TPP Zn

Z0036-1G Z0036-5G
TCZ0036-1GEA 485.06 CAD
TCZ0036-1G TCZ0036-5G
Zinc meso-tetraphenylporphine ≥97.0% (by HPLC)
Zinc meso-tetraphenylporphine
Formula: C₄₄H₂₈N₄Zn
MDL Number: MFCD00012155
CAS Number: 14074-80-7

Order Now

Specification Test Results

Appearance Green to Dark green to Dark blue powder to crystal
Purity (HPLC) min. 97.0 area%
Content of Zinc 8.6 to 10.6 %
Lambda max. 420.0 to 426.0 nm (THF)
Molar extinction coefficient min. 625000 (THF, 420.0 to 426.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR