Pyrilamine maleate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Mepyramine maleate, Anisopyradamine, Minihist, Paraminyl maleate, N1-(4-Methoxybenzyl)-N2,N2-dimethyl-N1-(pyridin-2-yl)ethane-1,2-diamine maleate, Diaminide maleate
P2369-25G
P2369-5G
TCP2369-25GEA
162.74
CAD
TCP2369-25G
TCP2369-5G
Pyrilamine maleate ≥98.0% (by HPLC, titration analysis)
Pyrilamine maleate
Formula:
C₂₁H₂₇N₃O₅ MW: 401.463 g/mol |
MDL Number:
MFCD00069333 CAS Number: 59-33-6 EINECS: 200-422-7 Merck Index: 14,07984 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 % (total of Ion-Pair) |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 100.0 to 104.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...