L-Glutamic acid dibenzyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D4839-25G
D4839-5G
TCD4839-25GEA
282.82
CAD
TCD4839-25G
TCD4839-5G
L-Glutamic acid dibenzyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
L-Glutamic acid dibenzyl ester hydrochloride
Formula:
C₁₉H₂₁NO₄·HCl |
MDL Number:
MFCD00035103 CAS Number: 4561-10-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 102.0 to 106.0 deg-C |
Specific rotation [a]20/D | +8.5 to +10.0 deg (C=2, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...